The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278625.1 | NP_001265554.1 | 8 - 9 | 1445 | 76 |
NM_001278626.1 | NP_001265555.1 | 8 - 9 | 1258 | 76 | |
NM_001278627.1 | NP_001265556.1 | 8 - 9 | 1168 | 76 | |
NM_001278628.1 | NP_001265557.1 | 7 - 8 | 1116 | 76 | |
NM_016652.5 | NP_057736.4 | 8 - 9 | 1481 | 76 | |
GenBank mRNA | AB016840.1 | - | 7 - 8 | 1078 | 76 |
AF111802.1 | - | 7 - 8 | 1072 | 76 | |
AF255443.2 | - | 8 - 9 | 1445 | 76 | |
AF318302.1 | - | 7 - 8 | 1063 | 76 | |
AF318303.1 | - | 8 - 9 | 1481 | 76 | |
AF318304.1 | - | 8 - 9 | 1258 | 76 | |
AF318305.1 | - | 8 - 9 | 1168 | 76 | |
AK022908.1 | - | 6 - 7 | 793 | 76 | |
AK023728.1 | - | 3 - 4 | 442 | 76 | |
AK292799.1 | - | 7 - 8 | 986 | 76 | |
AL832435.1 | - | 7 - 8 | 1090 | 76 |