Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDB3
Entrez Gene ID
11155
Gene Name
LIM domain binding 3
Gene Aliases
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location
Chr.10: 86664785 - 86736072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080114.1 NP_001073583.1 10 - 11 1550 60
NM_001171610.1 NP_001165081.1 11 - 12 2017 60
NM_007078.2 NP_009009.1 10 - 11 1880 60
XM_005269464.4 XP_005269521.1 11 - 12 2049 60
XM_005269466.4 XP_005269523.1 10 - 11 1859 60
XM_011539184.2 XP_011537486.1 13 - 14 2302 60
XM_011539185.2 XP_011537487.1 13 - 14 4196 60
XM_011539186.2 XP_011537488.1 12 - 13 2254 60
XM_011539187.2 XP_011537489.1 12 - 13 2112 60
XM_011539188.2 XP_011537490.1 12 - 13 2098 60
XM_011539189.1 XP_011537491.1 13 - 14 1851 60
XM_011539190.1 XP_011537492.1 11 - 12 1739 60
XM_011539191.2 XP_011537493.1 13 - 14 1767 60
XM_011539193.1 XP_011537495.1 11 - 12 1624 60
XM_011539194.1 XP_011537496.1 10 - 11 1435 60
XM_017015606.1 XP_016871095.1 12 - 13 3992 60
XM_017015607.1 XP_016871096.1 8 - 9 2228 60
GenBank mRNA AB014513.1 - 10 - 11 1878 60
AJ133767.1 - 10 - 11 1527 60
AJ133768.1 - 7 - 8 1086 60
AK294696.1 - 11 - 12 1700 60
AK304760.1 - 11 - 12 1959 60
KF772970.1 - 9 - 10 1668 60

View Full Product Details