The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001853.3 | NP_001844.3 | 22 - 23 | 1165 | 91 |
XM_011528545.1 | XP_011526847.1 | 22 - 23 | 1163 | 91 | |
XM_017027666.1 | XP_016883155.1 | 22 - 23 | 1163 | 91 | |
GenBank mRNA | BC007574.1 | - | 15 - 16 | 795 | 91 |
BC011705.2 | - | 22 - 23 | 1165 | 91 | |
L41162.1 | - | 22 - 23 | 1162 | 91 |