The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001958.3 | NP_001949.1 | 1 - 2 | 254 | 154 |
GenBank mRNA | BC110409.1 | - | 1 - 2 | 223 | 154 |
BM476361.1 | - | 1 - 2 | 265 | 154 |