Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL20A1
Entrez Gene ID
57642
Gene Name
collagen type XX alpha 1 chain
Gene Aliases
bA261N11.4
Chromosome Location
Chr.20: 63293169 - 63334847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020882.2 NP_065933.2 14 - 15 1906 88
XM_011528937.1 XP_011527239.1 14 - 15 1926 88
XM_011528938.1 XP_011527240.1 14 - 15 1926 88
XM_011528939.1 XP_011527241.1 14 - 15 1947 88
XM_011528940.1 XP_011527242.1 14 - 15 1944 88
GenBank mRNA AB040943.1 - 10 - 11 1358 88
BC043183.1 - 14 - 15 1911 88

View Full Product Details