The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098725.1 | NP_001092195.1 | 1 - 2 | 243 | 91 |
NM_021966.2 | NP_068801.1 | 1 - 2 | 243 | 91 | |
XM_017021676.1 | XP_016877165.1 | 1 - 2 | 199 | 91 | |
XM_017021677.1 | XP_016877166.1 | 1 - 2 | 199 | 91 | |
GenBank mRNA | BC003574.1 | - | 1 - 2 | 155 | 91 |
BC005831.2 | - | 1 - 2 | 155 | 91 | |
BC014024.1 | - | 1 - 2 | 159 | 91 | |
BG718395.1 | - | 1 - 2 | 248 | 91 | |
CR456796.1 | - | 1 - 2 | 113 | 91 | |
X82240.1 | - | 1 - 2 | 158 | 91 |