The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002744.4 | NP_002735.3 | 3 - 4 | 438 | 109 |
XM_011541773.1 | XP_011540075.1 | 3 - 4 | 434 | 109 | |
XM_011541775.2 | XP_011540077.1 | 3 - 4 | 434 | 109 | |
XM_017001789.1 | XP_016857278.1 | 3 - 4 | 434 | 109 | |
XM_017001790.1 | XP_016857279.1 | 3 - 4 | 434 | 109 | |
XM_017001803.1 | XP_016857292.1 | 3 - 4 | 434 | 109 | |
GenBank mRNA | AK290995.1 | - | 3 - 4 | 277 | 109 |
BC008058.2 | - | 3 - 4 | 385 | 109 | |
BC014270.2 | - | 3 - 4 | 438 | 109 | |
BT007082.1 | - | 3 - 4 | 277 | 109 | |
L14283.1 | - | 3 - 4 | 277 | 109 | |
Z15108.1 | - | 3 - 4 | 259 | 109 |