Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR3
Entrez Gene ID
55027
Gene Name
HEAT repeat containing 3
Gene Aliases
SYO1
Chromosome Location
Chr.16: 50065970 - 50107268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182922.2 NP_891552.1 13 - 14 1907 75
XM_005256013.2 XP_005256070.1 12 - 13 1722 75
XM_011523188.2 XP_011521490.1 13 - 14 1797 75
XM_017023385.1 XP_016878874.1 11 - 12 1860 75
XM_017023386.1 XP_016878875.1 11 - 12 1665 75
GenBank mRNA AF462442.1 - 10 - 11 2099 75
AK000725.1 - 12 - 13 1714 75
AK289949.1 - 13 - 14 1872 75
BC014415.1 - - 1884 75
BC018730.1 - 13 - 14 1872 75
BC033077.1 - 13 - 14 1773 75

View Full Product Details