Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR3
Entrez Gene ID
55027
Gene Name
HEAT repeat containing 3
Gene Aliases
SYO1
Chromosome Location
Chr.16: 50065970 - 50107268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182922.2 NP_891552.1 10 - 11 1537 84
XM_005256013.2 XP_005256070.1 9 - 10 1352 84
XM_011523188.2 XP_011521490.1 10 - 11 1427 84
XM_017023385.1 XP_016878874.1 8 - 9 1490 84
XM_017023386.1 XP_016878875.1 8 - 9 1295 84
GenBank mRNA AF462442.1 - 7 - 8 1729 84
AK000725.1 - 9 - 10 1344 84
AK289949.1 - 10 - 11 1502 84
BC014415.1 - - 1514 84
BC018730.1 - 10 - 11 1502 84
BC033077.1 - 10 - 11 1403 84

View Full Product Details