Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Chromosome Location
Chr.12: 56080025 - 56103507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001982.3 NP_001973.2 16 - 17 2190 80
GenBank mRNA AK291681.1 - 16 - 17 2043 80
AK294719.1 - 16 - 17 2047 80
AK295650.1 - 17 - 18 1957 80
AK300909.1 - 3 - 4 289 80
BC041579.1 - 4 - 5 366 80
BC082992.1 - 16 - 17 2029 80
M29366.1 - 16 - 17 2013 80
M34309.1 - 16 - 17 2112 80

View Full Product Details