The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001982.3 | NP_001973.2 | 27 - 28 | 3783 | 81 |
GenBank mRNA | AK124710.1 | - | 10 - 11 | 1965 | 81 |
AK125028.1 | - | 5 - 6 | 1325 | 81 | |
AK294719.1 | - | 26 - 27 | 3823 | 81 | |
AK295650.1 | - | 28 - 29 | 3550 | 81 | |
AK300909.1 | - | 14 - 15 | 1882 | 81 | |
BC002706.2 | - | 10 - 11 | 1454 | 81 | |
BC041579.1 | - | 12 - 13 | 1428 | 81 | |
BC082992.1 | - | 27 - 28 | 3622 | 81 | |
M34309.1 | - | 27 - 28 | 3705 | 81 |