Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Chromosome Location
Chr.12: 56080025 - 56103507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001982.3 NP_001973.2 4 - 5 820 67
GenBank mRNA AK291681.1 - 4 - 5 673 67
AK294719.1 - 4 - 5 677 67
AK295650.1 - 5 - 6 587 67
BC002706.2 - 4 - 5 640 67
BC082992.1 - 4 - 5 659 67
BT007226.1 - 4 - 5 544 67
M29366.1 - 4 - 5 643 67
M34309.1 - 4 - 5 742 67

View Full Product Details