The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001982.3 | NP_001973.2 | 8 - 9 | 1270 | 93 |
GenBank mRNA | AK291681.1 | - | 8 - 9 | 1123 | 93 |
AK294719.1 | - | 8 - 9 | 1127 | 93 | |
AK295650.1 | - | 9 - 10 | 1037 | 93 | |
BC082992.1 | - | 8 - 9 | 1109 | 93 | |
M29366.1 | - | 8 - 9 | 1093 | 93 | |
M34309.1 | - | 8 - 9 | 1192 | 93 |