The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152323.1 | NP_689536.1 | 3 - 4 | 235 | 126 |
XM_005268646.3 | XP_005268703.1 | 4 - 5 | 513 | 126 | |
XM_006719239.3 | XP_006719302.1 | 4 - 5 | 479 | 126 | |
GenBank mRNA | AF518404.1 | - | 4 - 5 | 368 | 126 |
BC032317.1 | - | 3 - 4 | 235 | 126 |