Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP90B2P
Entrez Gene ID
7190
Gene Name
heat shock protein 90 beta family member 2, pseudogene
Gene Aliases
GRP94P1, GRP94b, HSP, HSPCP2, TRA1P1, TRAP1
Chromosome Location
Chr.15: 99257525 - 99260276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073383.1 1 - 1 487 136

View Full Product Details