The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013358.2 | NP_037490.2 | 11 - 12 | 1404 | 65 |
XM_017001101.1 | XP_016856590.1 | 11 - 12 | 1307 | 65 | |
XM_017001102.1 | XP_016856591.1 | 11 - 12 | 1307 | 65 | |
XM_017001103.1 | XP_016856592.1 | 11 - 12 | 1407 | 65 | |
GenBank mRNA | AB033768.2 | - | 11 - 12 | 1395 | 65 |
AK026652.1 | - | 11 - 12 | 1406 | 65 | |
AK225541.1 | - | 11 - 12 | 1406 | 65 | |
AK314644.1 | - | 11 - 12 | 1405 | 65 | |
BC130574.1 | - | 11 - 12 | 1383 | 65 | |
BC136402.1 | - | 11 - 12 | 1383 | 65 |