The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013358.2 | NP_037490.2 | 14 - 15 | 1727 | 62 |
XM_017001101.1 | XP_016856590.1 | 14 - 15 | 1630 | 62 | |
XM_017001102.1 | XP_016856591.1 | 14 - 15 | 1630 | 62 | |
GenBank mRNA | AB033768.2 | - | 14 - 15 | 1718 | 62 |
AK026652.1 | - | 14 - 15 | 1729 | 62 | |
AK225541.1 | - | 14 - 15 | 1729 | 62 | |
AK293275.1 | - | 4 - 5 | 481 | 62 | |
AK298892.1 | - | 3 - 4 | 261 | 62 | |
AK314644.1 | - | 14 - 15 | 1728 | 62 | |
BC130574.1 | - | 14 - 15 | 1706 | 62 | |
BC136402.1 | - | 14 - 15 | 1706 | 62 |