The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013358.2 | NP_037490.2 | 4 - 5 | 505 | 82 |
XM_011541307.1 | XP_011539609.1 | 4 - 5 | 508 | 82 | |
XM_017001101.1 | XP_016856590.1 | 4 - 5 | 408 | 82 | |
XM_017001102.1 | XP_016856591.1 | 4 - 5 | 408 | 82 | |
XM_017001103.1 | XP_016856592.1 | 4 - 5 | 508 | 82 | |
GenBank mRNA | AB033768.2 | - | 4 - 5 | 496 | 82 |
AK026652.1 | - | 4 - 5 | 507 | 82 | |
AK225541.1 | - | 4 - 5 | 507 | 82 | |
AK314644.1 | - | 4 - 5 | 506 | 82 | |
BC130574.1 | - | 4 - 5 | 484 | 82 | |
BC136402.1 | - | 4 - 5 | 484 | 82 |