The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032414.2 | NP_115790.1 | 1 - 2 | 123 | 70 |
GenBank mRNA | AF333024.1 | - | 1 - 2 | 126 | 70 |
AY029225.1 | - | 1 - 2 | 120 | 70 | |
AY358683.1 | - | 1 - 2 | 162 | 70 | |
BC025399.1 | - | 1 - 2 | 100 | 70 | |
BM544315.1 | - | 1 - 2 | 149 | 70 |