The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003654.2 | 2 - 3 | 909 | 70 | |
NR_004859.1 | 3 - 4 | 1467 | 70 | ||
GenBank mRNA | AA280726.1 | - | 2 - 3 | 171 | 70 |
AF229246.1 | - | 3 - 4 | 2727 | 70 | |
AI218957.1 | - | 2 - 3 | 171 | 70 | |
AK022844.1 | - | 2 - 3 | 909 | 70 | |
AK290489.1 | - | 2 - 3 | 919 | 70 | |
AL109690.1 | - | 2 - 3 | 1467 | 70 | |
AW341587.1 | - | 3 - 4 | 251 | 70 | |
BC011547.1 | - | 2 - 3 | 891 | 70 | |
BC012929.1 | - | 2 - 3 | 891 | 70 | |
BC063836.1 | - | 2 - 3 | 865 | 70 |