The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206996.2 | NP_996879.1 | 1 - 2 | 148 | 70 |
XM_006710426.3 | XP_006710489.1 | 1 - 2 | 148 | 70 | |
XM_006710427.3 | XP_006710490.1 | 1 - 2 | 148 | 70 | |
XM_011540934.1 | XP_011539236.1 | 1 - 2 | 148 | 70 | |
GenBank mRNA | AK091816.1 | - | 1 - 2 | 134 | 70 |
AY555274.1 | - | 1 - 2 | 134 | 70 | |
BX648110.1 | - | 1 - 2 | 149 | 70 | |
DB060943.1 | - | 1 - 2 | 148 | 70 |