The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206996.2 | NP_996879.1 | 8 - 9 | 1244 | 87 |
XM_006710426.3 | XP_006710489.1 | 8 - 9 | 1244 | 87 | |
XM_006710427.3 | XP_006710490.1 | 7 - 8 | 1049 | 87 | |
XM_011540934.1 | XP_011539236.1 | 8 - 9 | 1244 | 87 | |
XM_011540935.2 | XP_011539237.1 | 7 - 8 | 1083 | 87 | |
GenBank mRNA | AK091816.1 | - | 8 - 9 | 1230 | 87 |
AY555274.1 | - | 8 - 9 | 1230 | 87 | |
BX648110.1 | - | 8 - 9 | 1245 | 87 |