The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247987.1 | NP_001234916.1 | 5 - 6 | 887 | 63 |
NM_020826.2 | NP_065877.1 | 3 - 4 | 669 | 63 | |
GenBank mRNA | AB037848.1 | - | 3 - 4 | 655 | 63 |
AJ303362.1 | - | 1 - 2 | 96 | 63 | |
AK291009.1 | - | 3 - 4 | 590 | 63 | |
AK308841.1 | - | 5 - 6 | 621 | 63 | |
AL512743.1 | - | 3 - 4 | 592 | 63 | |
BC093830.1 | - | 3 - 4 | 589 | 63 | |
BC093832.1 | - | 3 - 4 | 591 | 63 |