The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163147.1 | NP_001156619.1 | 9 - 10 | 1414 | 75 |
NM_001163148.1 | NP_001156620.1 | 10 - 11 | 1222 | 75 | |
NM_001163149.1 | NP_001156621.1 | 9 - 10 | 1093 | 75 | |
NM_001163150.1 | NP_001156622.1 | 7 - 8 | 1159 | 75 | |
NM_001163151.1 | NP_001156623.1 | 6 - 7 | 1105 | 75 | |
NM_001163152.1 | NP_001156624.1 | 6 - 7 | 970 | 75 | |
NM_004956.4 | NP_004947.2 | 11 - 12 | 1680 | 75 | |
XM_005249635.4 | XP_005249692.1 | 10 - 11 | 1193 | 75 | |
XM_005249636.4 | XP_005249693.1 | 10 - 11 | 1116 | 75 | |
XM_011515167.2 | XP_011513469.1 | 9 - 10 | 1100 | 75 | |
XM_011515168.2 | XP_011513470.1 | 10 - 11 | 1126 | 75 | |
XM_011515170.2 | XP_011513472.1 | 9 - 10 | 978 | 75 | |
GenBank mRNA | AB209202.1 | - | 9 - 10 | 1061 | 75 |
AK293802.1 | - | 6 - 7 | 658 | 75 | |
AK294755.1 | - | 9 - 10 | 1061 | 75 | |
AK299693.1 | - | 6 - 7 | 1043 | 75 | |
AK309190.1 | - | 7 - 8 | 895 | 75 | |
AK311134.1 | - | 6 - 7 | 837 | 75 | |
AK311194.1 | - | 7 - 8 | 891 | 75 | |
AK312863.1 | - | 11 - 12 | 1362 | 75 | |
AK316007.1 | - | 6 - 7 | 897 | 75 | |
BC098403.1 | - | 11 - 12 | 1354 | 75 | |
BC106762.1 | - | 10 - 11 | 1173 | 75 | |
BC106763.1 | - | 10 - 11 | 1172 | 75 | |
CR627389.1 | - | 7 - 8 | 914 | 75 | |
U17163.1 | - | 9 - 10 | 941 | 75 | |
X87175.1 | - | 9 - 10 | 1102 | 75 |