The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015431.3 | NP_056246.3 | 2 - 3 | 564 | 69 |
GenBank mRNA | AF327057.1 | - | 2 - 3 | 564 | 69 |
AK096188.1 | - | 2 - 3 | 446 | 69 | |
BC074748.2 | - | 1 - 2 | 108 | 69 | |
BX644877.1 | - | 2 - 3 | 120 | 69 | |
DN998591.1 | - | 2 - 3 | 563 | 69 |