The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113178.2 | NP_001106649.1 | 14 - 15 | 4983 | 88 |
NM_006599.3 | NP_006590.1 | 13 - 14 | 4932 | 88 | |
NM_138713.3 | NP_619727.2 | 14 - 15 | 4986 | 88 | |
NM_138714.3 | NP_619728.2 | 15 - 16 | 5048 | 88 | |
NM_173214.2 | NP_775321.1 | 14 - 15 | 4994 | 88 | |
XM_006721125.3 | XP_006721188.1 | 13 - 14 | 5657 | 88 | |
XM_011522817.2 | XP_011521119.1 | 13 - 14 | 4929 | 88 | |
XM_011522818.2 | XP_011521120.1 | 16 - 17 | 5121 | 88 | |
XM_011522819.2 | XP_011521121.1 | 16 - 17 | 4703 | 88 | |
XM_011522820.2 | XP_011521122.1 | 13 - 14 | 4133 | 88 | |
XM_011522822.2 | XP_011521124.1 | 12 - 13 | 4081 | 88 | |
XM_017022870.1 | XP_016878359.1 | 13 - 14 | 5281 | 88 | |
XM_017022871.1 | XP_016878360.1 | 14 - 15 | 5005 | 88 | |
XM_017022872.1 | XP_016878361.1 | 13 - 14 | 4862 | 88 | |
XM_017022873.1 | XP_016878362.1 | 13 - 14 | 4471 | 88 | |
GenBank mRNA | AB020634.1 | - | 13 - 14 | 4914 | 88 |
AF089824.1 | - | 14 - 15 | 4936 | 88 | |
AF346509.1 | - | 15 - 16 | 5934 | 88 | |
AJ005684.1 | - | 14 - 15 | 4610 | 88 | |
AJ243298.1 | - | 14 - 15 | 4753 | 88 | |
AJ243299.1 | - | 14 - 15 | 4745 | 88 | |
BC131509.1 | - | 14 - 15 | 4887 | 88 | |
BC146765.1 | - | 13 - 14 | 4914 | 88 | |
U80231.1 | - | 2 - 3 | 1627 | 88 |