The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019065.2 | NP_061938.2 | 8 - 9 | 813 | 85 |
XM_011523135.1 | XP_011521437.1 | 7 - 8 | 630 | 85 | |
XM_017023317.1 | XP_016878806.1 | 6 - 7 | 1668 | 85 | |
GenBank mRNA | AF070637.1 | - | 4 - 5 | 408 | 85 |
AF193758.1 | - | 4 - 5 | 406 | 85 | |
AK127376.1 | - | 8 - 9 | 734 | 85 | |
AY299331.1 | - | 8 - 9 | 813 | 85 | |
BC016979.1 | - | 6 - 7 | 809 | 85 | |
BC131615.1 | - | 3 - 4 | 299 | 85 |