Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL30
Entrez Gene ID
377007
Gene Name
kelch like family member 30
Gene Aliases
-
Chromosome Location
Chr.2: 238138722 - 238152906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198582.3 NP_940984.3 4 - 5 1107 64
GenBank mRNA AK125364.1 - 3 - 4 979 64
BC103703.1 - 3 - 4 977 64
BC104749.1 - 3 - 4 977 64
BC105696.1 - 3 - 4 977 64

View Full Product Details