Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MKNK1
- Entrez Gene ID
-
8569
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
- Gene Aliases
-
MNK1
- Chromosome Location
-
Chr.1: 46557407 - 46607140 on Build GRCh38
View Full Product Details