Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
Chromosome Location
Chr.12: 122834498 - 122862961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003959.2 NP_003950.1 20 - 21 2161 70
XM_011538963.1 XP_011537265.1 20 - 21 2464 70
GenBank mRNA AB013384.1 - 14 - 15 1514 70
AB014555.1 - 20 - 21 2095 70

View Full Product Details