Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD7
Entrez Gene ID
149951
Gene Name
COMM domain containing 7
Gene Aliases
C20orf92, dJ1085F17.3
Chromosome Location
Chr.20: 32702691 - 32744008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099339.1 NP_001092809.1 2 - 3 757 78
NM_053041.2 NP_444269.2 2 - 3 760 78
XM_005260299.3 XP_005260356.1 2 - 3 747 78
XM_005260300.3 XP_005260357.1 2 - 3 744 78
XM_011528604.2 XP_011526906.1 2 - 3 747 78
XM_017027685.1 XP_016883174.1 2 - 3 744 78
XM_017027686.1 XP_016883175.1 2 - 3 747 78
XM_017027687.1 XP_016883176.1 2 - 3 747 78
GenBank mRNA AK027893.1 - 2 - 3 207 78
AK096307.1 - 2 - 3 749 78
AK097059.1 - 3 - 4 284 78
AY542162.1 - 2 - 3 78
BC000628.2 - 2 - 3 193 78
BC047440.1 - 2 - 3 78
DN993528.1 - 2 - 3 504 78

View Full Product Details