Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNB
Entrez Gene ID
167838
Gene Name
taxilin beta
Gene Aliases
C6orf198, LST001, MDP77, dJ522B19.2
Chromosome Location
Chr.6: 139159160 - 139324011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153235.3 NP_694967.3 4 - 5 851 87
XM_005266836.1 XP_005266893.1 5 - 6 1218 87
XM_011535505.2 XP_011533807.1 4 - 5 955 87
XM_011535506.2 XP_011533808.1 3 - 4 401 87
XM_017010322.1 XP_016865811.1 5 - 6 1081 87
GenBank mRNA AB085905.1 - 4 - 5 851 87
AK314725.1 - 4 - 5 799 87
AL831988.1 - 4 - 5 789 87
AL832322.1 - 1 - 2 149 87
AL834248.2 - 4 - 5 779 87
BC115383.1 - 4 - 5 806 87
BC115384.2 - 4 - 5 806 87
BX647477.1 - 4 - 5 799 87
BX647633.1 - 4 - 5 777 87

View Full Product Details