Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNB
Entrez Gene ID
167838
Gene Name
taxilin beta
Gene Aliases
C6orf198, LST001, MDP77, dJ522B19.2
Chromosome Location
Chr.6: 139159160 - 139324011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153235.3 NP_694967.3 8 - 9 1342 70
XM_005266836.1 XP_005266893.1 9 - 10 1709 70
XM_011535505.2 XP_011533807.1 8 - 9 1446 70
XM_011535506.2 XP_011533808.1 7 - 8 892 70
XM_017010322.1 XP_016865811.1 9 - 10 1572 70
GenBank mRNA AB085905.1 - 8 - 9 1342 70
AK314725.1 - 8 - 9 1290 70
AL831988.1 - 8 - 9 1280 70
AL832322.1 - 5 - 6 640 70
AL834248.2 - 8 - 9 1270 70
BC115383.1 - 8 - 9 1297 70
BC115384.2 - 8 - 9 1297 70
BX647477.1 - 8 - 9 1290 70
BX647633.1 - 8 - 9 1268 70

View Full Product Details