The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020844.2 | NP_065895.2 | 3 - 4 | 653 | 74 |
XM_005273584.4 | XP_005273641.1 | 3 - 4 | 664 | 74 | |
XM_005273585.4 | XP_005273642.1 | 4 - 5 | 675 | 74 | |
XM_005273586.4 | XP_005273643.1 | 4 - 5 | 709 | 74 | |
XM_011544596.2 | XP_011542898.1 | 3 - 4 | 1124 | 74 | |
XM_011544597.2 | XP_011542899.1 | 3 - 4 | 476 | 74 | |
XM_011544598.2 | XP_011542900.1 | 3 - 4 | 745 | 74 | |
XM_011544600.2 | XP_011542902.1 | 3 - 4 | 483 | 74 | |
XM_011544601.2 | XP_011542903.1 | 3 - 4 | 606 | 74 | |
XM_017013705.1 | XP_016869194.1 | 3 - 4 | 514 | 74 | |
XM_017013706.1 | XP_016869195.1 | 3 - 4 | 735 | 74 | |
XM_017013707.1 | XP_016869196.1 | 4 - 5 | 880 | 74 | |
XM_017013713.1 | XP_016869202.1 | 3 - 4 | 779 | 161 | |
GenBank mRNA | AK296010.1 | - | 4 - 5 | 506 | 74 |