The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251987.1 | NP_001238916.1 | 19 - 20 | 1594 | 89 |
NM_006073.3 | NP_006064.2 | 19 - 20 | 1591 | 89 | |
GenBank mRNA | AJ489257.2 | - | 19 - 20 | 1489 | 89 |
CR749591.1 | - | 9 - 10 | 350 | 89 | |
U18985.1 | - | 19 - 20 | 1295 | 89 |