Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRDN
Entrez Gene ID
10345
Gene Name
triadin
Gene Aliases
CPVT5, TDN, TRISK
Chromosome Location
Chr.6: 123216339 - 123637093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006073.3 NP_006064.2 23 - 24 1789 80
GenBank mRNA CR749591.1 - 12 - 13 497 80
U18985.1 - 23 - 24 1493 80

View Full Product Details