The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008539.3 | NP_001008539.3 | 9 - 10 | 1679 | 78 |
NM_001164771.1 | NP_001158243.1 | 9 - 10 | 1675 | 78 | |
NM_003046.5 | NP_003037.4 | 9 - 10 | 1672 | 78 | |
XM_005273610.4 | XP_005273667.1 | 10 - 11 | 1740 | 78 | |
XM_005273611.4 | XP_005273668.1 | 10 - 11 | 1643 | 78 | |
XM_005273612.4 | XP_005273669.1 | 10 - 11 | 1737 | 78 | |
XM_017013746.1 | XP_016869235.1 | 9 - 10 | 1678 | 78 | |
XM_017013747.1 | XP_016869236.1 | 10 - 11 | 1737 | 78 | |
GenBank mRNA | AK314287.1 | - | 9 - 10 | 1677 | 78 |
AL832016.2 | - | 4 - 5 | 568 | 78 | |
BC069648.1 | - | 9 - 10 | 1582 | 78 | |
BC104905.1 | - | 9 - 10 | 1565 | 78 | |
BC113661.1 | - | 9 - 10 | 1565 | 78 | |
BC143583.1 | - | 9 - 10 | 1568 | 78 | |
D29990.1 | - | 8 - 9 | 1507 | 78 | |
U76368.1 | - | 9 - 10 | 1698 | 78 |