Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR7
Entrez Gene ID
9108
Gene Name
myotubularin related protein 7
Gene Aliases
-
Chromosome Location
Chr.8: 17296794 - 17413531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004686.4 NP_004677.3 1 - 2 262 108
XM_017013956.1 XP_016869445.1 1 - 2 84 108
GenBank mRNA AK316283.1 - 1 - 2 108
BC130577.1 - 1 - 2 108
BC130579.1 - 1 - 2 108
CR749240.1 - 1 - 2 108
DC325464.1 - 1 - 2 108

View Full Product Details