The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286658.2 | NP_001273587.1 | 3 - 4 | 1565 | 127 |
NM_001286659.2 | NP_001273588.1 | 3 - 4 | 1565 | 127 | |
NM_014832.4 | NP_055647.2 | 3 - 4 | 1565 | 127 | |
XM_005266603.1 | XP_005266660.1 | 3 - 4 | 2043 | 127 | |
XM_005266605.2 | XP_005266662.1 | 3 - 4 | 893 | 127 | |
XM_006719903.3 | XP_006719966.1 | 3 - 4 | 1253 | 127 | |
XM_011535331.1 | XP_011533633.1 | 3 - 4 | 2043 | 127 | |
XM_017020882.1 | XP_016876371.1 | 3 - 4 | 774 | 127 | |
XM_017020883.1 | XP_016876372.1 | 3 - 4 | 879 | 127 | |
GenBank mRNA | AB011175.1 | - | 3 - 4 | 1511 | 127 |
AB449885.1 | - | 3 - 4 | 1164 | 127 | |
AK307493.1 | - | 3 - 4 | 1565 | 127 | |
BC151239.1 | - | 3 - 4 | 1503 | 127 | |
FM207106.1 | - | 3 - 4 | 1164 | 127 | |
FM207107.1 | - | 3 - 4 | 1164 | 127 |