Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
Chromosome Location
Chr.13: 75284664 - 75482646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286658.2 NP_001273587.1 5 - 6 1816 76
NM_001286659.2 NP_001273588.1 5 - 6 1816 76
NM_014832.4 NP_055647.2 5 - 6 1816 76
XM_005266603.1 XP_005266660.1 5 - 6 2294 76
XM_005266605.2 XP_005266662.1 5 - 6 1144 76
XM_006719903.3 XP_006719966.1 5 - 6 1504 76
XM_011535331.1 XP_011533633.1 5 - 6 2294 76
XM_017020882.1 XP_016876371.1 5 - 6 1025 76
XM_017020883.1 XP_016876372.1 5 - 6 1130 76
GenBank mRNA AB011175.1 - 5 - 6 1762 76
AB449885.1 - 5 - 6 1415 76
AK307493.1 - 5 - 6 1816 76
BC151239.1 - 5 - 6 1754 76
FM207106.1 - 5 - 6 1415 76
FM207107.1 - 5 - 6 1415 76

View Full Product Details