The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145400.1 | NP_001138872.1 | 7 - 8 | 1399 | 67 |
NM_139174.3 | NP_631913.3 | 8 - 9 | 1645 | 67 | |
XM_005255814.1 | XP_005255871.1 | 7 - 8 | 1429 | 67 | |
XM_011522919.1 | XP_011521221.1 | 8 - 9 | 1615 | 67 | |
XM_011522920.1 | XP_011521222.1 | 8 - 9 | 1340 | 67 | |
GenBank mRNA | AF447586.1 | - | 7 - 8 | 1337 | 67 |
AK093046.1 | - | 8 - 9 | 1624 | 67 | |
AK093523.1 | - | 5 - 6 | 1899 | 67 | |
AK315171.1 | - | 7 - 8 | 1318 | 67 | |
BC033491.1 | - | 7 - 8 | 1337 | 67 |