The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199865.1 | NP_001186794.1 | 6 - 7 | 711 | 93 |
NM_001199866.1 | NP_001186795.1 | 6 - 7 | 711 | 93 | |
NM_024704.4 | NP_078980.3 | 6 - 7 | 711 | 93 | |
XM_005260750.3 | XP_005260807.1 | 6 - 7 | 690 | 93 | |
XM_005260751.4 | XP_005260808.1 | 6 - 7 | 706 | 93 | |
XM_005260752.3 | XP_005260809.1 | 6 - 7 | 690 | 93 | |
XM_005260753.3 | XP_005260810.1 | 6 - 7 | 688 | 93 | |
XM_005260754.3 | XP_005260811.1 | 6 - 7 | 688 | 93 | |
XM_005260755.3 | XP_005260812.1 | 6 - 7 | 734 | 93 | |
XM_006723588.3 | XP_006723651.1 | 6 - 7 | 688 | 93 | |
XM_017027926.1 | XP_016883415.1 | 6 - 7 | 706 | 93 | |
GenBank mRNA | AB046810.2 | - | 6 - 7 | 628 | 93 |
AK095322.1 | - | 6 - 7 | 648 | 93 | |
AK307399.1 | - | 6 - 7 | 594 | 93 | |
AY166853.1 | - | 6 - 7 | 710 | 93 | |
BC050577.1 | - | 6 - 7 | 711 | 93 | |
BC150261.1 | - | 6 - 7 | 628 | 93 | |
BX647572.1 | - | 6 - 7 | 630 | 93 |