The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206927.1 | NP_001193856.1 | 41 - 42 | 5847 | 90 |
XM_011514318.2 | XP_011512620.1 | 40 - 41 | 5795 | 90 | |
XM_011514319.2 | XP_011512621.1 | 41 - 42 | 5858 | 90 | |
XM_011514320.2 | XP_011512622.1 | 39 - 40 | 5621 | 90 | |
XM_011514321.1 | XP_011512623.1 | 38 - 39 | 5205 | 90 | |
XM_017010325.1 | XP_016865814.1 | 41 - 42 | 5858 | 90 | |
XM_017010326.1 | XP_016865815.1 | 41 - 42 | 5858 | 90 | |
XM_017010327.1 | XP_016865816.1 | 41 - 42 | 5859 | 90 | |
GenBank mRNA | AF356519.1 | - | 39 - 40 | 5341 | 90 |