The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206927.1 | NP_001193856.1 | 6 - 7 | 1063 | 153 |
XM_011514318.2 | XP_011512620.1 | 6 - 7 | 1074 | 153 | |
XM_011514319.2 | XP_011512621.1 | 6 - 7 | 1074 | 153 | |
XM_011514320.2 | XP_011512622.1 | 4 - 5 | 837 | 153 | |
XM_011514321.1 | XP_011512623.1 | 3 - 4 | 421 | 153 | |
XM_017010325.1 | XP_016865814.1 | 6 - 7 | 1074 | 153 | |
XM_017010326.1 | XP_016865815.1 | 6 - 7 | 1074 | 153 | |
XM_017010327.1 | XP_016865816.1 | 6 - 7 | 1075 | 153 | |
GenBank mRNA | AF356519.1 | - | 4 - 5 | 557 | 153 |