The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206927.1 | NP_001193856.1 | 75 - 76 | 11386 | 159 |
XM_011514318.2 | XP_011512620.1 | 74 - 75 | 11334 | 159 | |
XM_011514319.2 | XP_011512621.1 | 74 - 75 | 11289 | 159 | |
XM_011514320.2 | XP_011512622.1 | 73 - 74 | 11160 | 159 | |
XM_011514321.1 | XP_011512623.1 | 72 - 73 | 10744 | 159 | |
XM_017010325.1 | XP_016865814.1 | 75 - 76 | 11397 | 159 | |
XM_017010326.1 | XP_016865815.1 | 75 - 76 | 11397 | 159 | |
GenBank mRNA | AF356519.1 | - | 73 - 74 | 10880 | 159 |
AK093653.1 | - | 1 - 2 | 108 | 159 | |
AW503176.1 | - | 2 - 3 | 238 | 159 |