The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175736.4 | NP_783863.4 | 22 - 23 | 2907 | 79 |
NM_198900.2 | NP_944489.2 | 21 - 22 | 2754 | 79 | |
XM_005269218.2 | XP_005269275.1 | 22 - 23 | 2886 | 79 | |
XM_011538968.2 | XP_011537270.1 | 22 - 23 | 3376 | 79 | |
XM_011538969.2 | XP_011537271.1 | 22 - 23 | 3375 | 79 | |
XM_011538970.2 | XP_011537272.1 | 22 - 23 | 3375 | 79 | |
XM_011538971.2 | XP_011537273.1 | 22 - 23 | 2951 | 79 | |
XM_011538972.2 | XP_011537274.1 | 21 - 22 | 3221 | 79 | |
XM_011538973.2 | XP_011537275.2 | 18 - 19 | 2366 | 79 | |
XM_011538974.2 | XP_011537276.1 | 23 - 24 | 2708 | 79 | |
GenBank mRNA | AB095934.2 | - | 22 - 23 | 2907 | 79 |
AK127200.1 | - | 9 - 10 | 1380 | 79 | |
AK128195.1 | - | 21 - 22 | 2734 | 79 | |
AL162062.1 | - | 6 - 7 | 864 | 79 | |
BC033181.1 | - | 6 - 7 | 691 | 79 | |
BC159100.1 | - | 22 - 23 | 2907 | 79 |