The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198976.2 | NP_945327.2 | 11 - 12 | 1449 | 125 |
GenBank mRNA | AF161479.1 | - | 11 - 12 | 123 | |
AJ238374.1 | - | 6 - 7 | 846 | 125 | |
AJ238375.1 | - | 6 - 7 | 772 | 125 | |
AJ238379.1 | - | 11 - 12 | 1383 | 125 | |
AK001316.1 | - | 11 - 12 | 1357 | 125 | |
AK023927.1 | - | 11 - 12 | 1374 | 125 | |
AK293410.1 | - | 11 - 12 | 1401 | 125 | |
BC014952.1 | - | 11 - 12 | 1385 | 125 | |
BX647417.1 | - | 11 - 12 | 1384 | 125 |