Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKIV2L
Entrez Gene ID
6499
Gene Name
Ski2 like RNA helicase
Gene Aliases
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, SKIV2L1, THES2
Chromosome Location
Chr.6: 31958804 - 31969852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006929.4 NP_008860.4 6 - 7 925 96
XM_011514815.2 XP_011513117.1 6 - 7 632 96
GenBank mRNA AB209605.1 - 6 - 7 581 96
AK293881.1 - 6 - 7 588 96
AK297230.1 - 4 - 5 297 96
AK303302.1 - 6 - 7 572 96
BC015758.1 - 6 - 7 538 96
U09877.1 - 6 - 7 647 96
Z48796.1 - 6 - 7 588 96

View Full Product Details