The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012327.5 | NP_036459.1 | 16 - 17 | 1952 | 82 |
NM_176787.4 | NP_789744.1 | 17 - 18 | 2029 | 82 | |
XM_011525889.1 | XP_011524191.1 | 17 - 18 | 1997 | 82 | |
XM_011525890.1 | XP_011524192.1 | 17 - 18 | 2194 | 82 | |
XM_011525891.1 | XP_011524193.1 | 17 - 18 | 1912 | 82 | |
XM_011525892.1 | XP_011524194.1 | 16 - 17 | 1934 | 82 | |
XM_011525893.1 | XP_011524195.1 | 16 - 17 | 1885 | 82 | |
XM_011525894.1 | XP_011524196.1 | 16 - 17 | 1796 | 82 | |
XM_011525895.1 | XP_011524197.1 | 15 - 16 | 1794 | 82 | |
XM_011525896.1 | XP_011524198.1 | 17 - 18 | 1997 | 82 | |
XM_011525898.1 | XP_011524200.1 | 17 - 18 | 1997 | 82 | |
GenBank mRNA | AF109219.1 | - | 16 - 17 | 1865 | 82 |
AK315134.1 | - | 17 - 18 | 1997 | 82 | |
AL137607.1 | - | 13 - 14 | 1264 | 82 | |
BC028363.2 | - | 17 - 18 | 1966 | 82 |