Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKI55
Entrez Gene ID
150967
Gene Name
DKFZp434H1419
Gene Aliases
-
Chromosome Location
Chr.2: 216216889 - 216220192 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037701.1 1 - 1 3249 94
GenBank mRNA AK125369.1 - 1 - 1 3093 94
AL137534.1 - 1 - 1 2557 94


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI123325.1 - 1 Chr. 2 - 216219771 - 216220184 [-] on Build GRCh38 94
GenBankmRNA AI524813.1 - 1 Chr. 2 - 216219691 - 216220192 [-] on Build GRCh38 94
GenBankmRNA AI025922.1 - 1 Chr. 2 - 216219816 - 216220192 [-] on Build GRCh38 94
GenBankmRNA AA032264.1 - 1 Chr. 2 - 216219536 - 216220184 [-] on Build GRCh38 94
GenBankmRNA AA255534.1 - 1 Chr. 2 - 216219787 - 216220192 [-] on Build GRCh38 94
GenBankmRNA BF476629.1 - 1 Chr. 2 - 216219758 - 216220192 [-] on Build GRCh38 94
GenBankmRNA DB325686.1 - 1 Chr. 2 - 216219623 - 216220190 [-] on Build GRCh38 96
GenBankmRNA AA902947.1 - 1 Chr. 2 - 216220043 - 216220192 [-] on Build GRCh38 94
GenBankmRNA BG150088.1 - 1 Chr. 2 - 216219752 - 216220192 [-] on Build GRCh38 94
GenBankmRNA DB573280.1 - 1 417 Chr. 2 - 216219721 - 216220192 [+] on Build GRCh38 94
GenBankmRNA AI375285.1 - 1 Chr. 2 - 216219770 - 216220185 [-] on Build GRCh38 94
GenBankmRNA DB575568.1 - 1 430 Chr. 2 - 216219708 - 216220192 [+] on Build GRCh38 94
GenBankmRNA AW291129.1 - 1 Chr. 2 - 216219831 - 216220186 [-] on Build GRCh38 94
GenBankmRNA AI371775.1 - 1 Chr. 2 - 216219731 - 216220185 [-] on Build GRCh38 94
GenBankmRNA AW293379.1 - 1 Chr. 2 - 216219713 - 216220185 [-] on Build GRCh38 94
GenBankmRNA W23761.1 - Chr. - - [-] on Build 94
GenBankmRNA AI202999.1 - 1 Chr. 2 - 216219839 - 216220186 [-] on Build GRCh38 94
GenBankmRNA AI280365.1 - 1 Chr. 2 - 216219794 - 216220188 [-] on Build GRCh38 94
GenBankmRNA DB563055.1 - 1 369 Chr. 2 - 216219770 - 216220192 [+] on Build GRCh38 94
GenBankmRNA CA312922.1 - 1 Chr. 2 - 216219713 - 216220192 [-] on Build GRCh38 94
GenBankmRNA DR977897.1 - 1 78 Chr. 2 - 216220059 - 216220185 [+] on Build GRCh38 94
GenBankmRNA AA069274.1 - 1 Chr. 2 - 216219870 - 216220192 [-] on Build GRCh38 94
GenBankmRNA AI827261.1 - 1 Chr. 2 - 216219773 - 216220185 [-] on Build GRCh38 94
GenBankmRNA AI702204.1 - 1 Chr. 2 - 216219835 - 216220192 [-] on Build GRCh38 94
GenBankmRNA AW470107.1 - 1 Chr. 2 - 216219686 - 216220186 [-] on Build GRCh38 94
GenBankmRNA BM669630.1 - 1 Chr. 2 - 216219708 - 216220192 [-] on Build GRCh38 94


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream X-ray repair cross complementing 5 XRCC5 KARP-1, KARP1, KU80, KUB2, Ku86, NFIV -13.5kb Anti-sense Chr. - 216109297 - 216206293 on Build GRCh38
Downstream membrane associated ring-CH-type finger 4 MARCH4 MARCH-IV, RNF174 37.7kb Sense Chr. - 216257862 - 216372027 on Build GRCh38

View Full Product Details